Learn More
N-Methylmaleimide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 127081000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Methylmaleimide | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD00005508 | |
21, 399 | |
SEEYREPSKCQBBF-UHFFFAOYSA-N | |
1-methylpyrrole-2,5-dione | |
70261 | |
97% |
930-88-1 | |
100.0 | |
96% min. (GC) | |
C5H5NO2 | |
100g | |
n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione | |
CN1C(=O)C=CC1=O | |
111.1 | |
111.1 |
Safety and Handling
GHS H Statement
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 213-226-1
RUO â Research Use Only