Please login to your online account to display your discounted pricing

N-Methylmaleimide, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  127080250

 View more versions of this product

Catalog No. AC127080250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystals
Formula Weight 111.1
Quantity 25g
Color Light yellow
Beilstein 21, 399
Melting Point 93° to 98°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C5H5NO2
Packaging Glass bottle
MDL Number MFCD00005508
CAS 930-88-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Chemical Name or Material N-Methylmaleimide
Assay Percent Range 97%

Specifications

Chemical Name or Material N-Methylmaleimide
CAS 930-88-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H5NO2
Formula Weight 111.1
MDL Number MFCD00005508
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

SDS & Safety Documents