Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylmaleamic acid, 97%

$52.59 - $188.35

Chemical Identifiers

Chemical Name or Material N-Methylmaleamic acid
CAS 6936-48-7
Molecular Formula C5H7NO3
Formula Weight 129.11
MDL Number MFCD00014012
Beilstein 1905722
Synonym Maleic acid monomethylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0061414

  SDS
alfa aesar™
L0061414
25g Each for $188.35

AAL0061406

  SDS
alfa aesar™
L0061406
5g Each for $52.59
Description & Specifications

Specifications

Chemical Name or Material N-Methylmaleamic acid
CAS 6936-48-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 154° to 157°C
Assay 97%
Quantity 25g
Molecular Formula C5H7NO3
Formula Weight 129.11
MDL Number MFCD00014012
Beilstein 1905722
Synonym Maleic acid monomethylamide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.