Please login to your online account to display your discounted pricing

N-Methyllidocaine iodide, Tocris

Enhances biosynthesis of phosphatidylinositol

$159.50 - $669.90

Chemical Identifiers

Chemical Name or Material N-Methyllidocaine iodide
Assay Percent Range >98%
Molecular Formula C15H25IN2O
Formula Weight 376.28
Synonym 2-[(2, 6-Dimethylphenyl)amino]-N, N-diethyl-N-methyl-2-oxoethanaminium iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

104250

 
r & d systems
RDS104250
50mg Each for $669.90

104210

 
r & d systems
RDS104210
10mg Each for $159.50
Description & Specifications

Specifications

Formula Weight 376.28
Quantity 50mg
Molecular Formula C15H25IN2O
CAS 1462-71-1
Assay Percent Range >98%
Synonym 2-[(2, 6-Dimethylphenyl)amino]-N, N-diethyl-N-methyl-2-oxoethanaminium iodide
Recommended Storage Desiccate at +4°C
Chemical Name or Material N-Methyllidocaine iodide
Safety and Handling

Desiccate at +4°C