missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyllidocaine iodide, Tocris

Enhances biosynthesis of phosphatidylinositol

Manufacturer:  R & D Systems RDS104210

 View more versions of this product

Catalog No. 104210


 

Specifications Description & Specifications

Specifications

N-Methyllidocaine iodide
>98%
376.28
10mg
1462-71-1
C15H25IN2O
2-[(2, 6-Dimethylphenyl)amino]-N, N-diethyl-N-methyl-2-oxoethanaminium iodide

Chemical Identifiers

>98%
376.28
N-Methyllidocaine iodide
C15H25IN2O
2-[(2, 6-Dimethylphenyl)amino]-N, N-diethyl-N-methyl-2-oxoethanaminium iodide

Safety and Handling

Recommended Storage : Desiccate at +4°C