Learn More
N-Methylisobutylamine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04294703
Description
N-Methylisobutylamine is used as an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Methylisobutylamine | |
0.73 g/mL | |
Amine-like | |
MFCD00015043 | |
UN2733 | |
Miscible with water. | |
CC(C)CNC | |
87.166 | |
87.16 | |
Liquid |
625-43-4 | |
78°C | |
C5H13N | |
1 g | |
n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin | |
QKYWADPCTHTJHQ-UHFFFAOYSA-N | |
N,2-dimethylpropan-1-amine | |
12249 | |
≥98% |
Chemical Identifiers
625-43-4 | |
87.166 | |
QKYWADPCTHTJHQ-UHFFFAOYSA-N | |
12249 | |
CC(C)CNC |
C5H13N | |
MFCD00015043 | |
n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin | |
N,2-dimethylpropan-1-amine |
Safety and Handling
GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 210-893-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only