Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylisatoic anhydride, tech. 90%

$70.81 - $70.81

Chemical Identifiers

Chemical Name or Material N-Methylisatoic anhydride
CAS 10328-92-4
Purity Grade Technical Grade
Molecular Formula C9H7NO3
Formula Weight 177.16
MDL Number MFCD00006815
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2409922

 
alfa aesar™
B2409922
100g Each for $70.81
Description & Specifications

Specifications

Formula Weight 177.16
Chemical Name or Material N-Methylisatoic anhydride
TSCA TSCA
Melting Point ≈160°C (dec.)
Purity Grade Technical Grade
Assay 90%
Quantity 100g
Molecular Formula C9H7NO3
MDL Number MFCD00006815
CAS 10328-92-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DM3140000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.