Please login to your online account to display your discounted pricing

N-Methylindole-3-boronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Methylindole-3-boronic Acid Pinacol Ester
CAS 683229-61-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C15H20BNO2
Formula Weight 257.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC441010010

  SDS
thermo fisher scientific
AC441010010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441010050

  SDS
thermo fisher scientific
AC441010050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Low Melting Solid
Assay ≥96.0% (GC)
Formula Weight 257.14
Quantity 1g
Color White to pale brown
Melting Point 105° to 113°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material N-Methylindole-3-boronic acid pinacol ester
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C15H20BNO2
CAS 683229-61-0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.