Please login to your online account to display your discounted pricing

Nα-Methylhistamine dihydrochloride, Tocris

Non-selective H3 agonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material Nα-Methylhistamine dihydrochloride
Assay Percent Range >98%
Molecular Formula C6H11N3·2HCl
Formula Weight 198.09
Synonym N-Methyl-1H-imidazole-4-ethanamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

057310

 
r & d systems
RDS057310
10mg Each for $168.15

057350

 
r & d systems
RDS057350
50mg Each for $697.40
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 198.09
Chemical Name or Material Nα-Methylhistamine dihydrochloride
Assay Percent Range >98%
Molecular Formula C6H11N3·2HCl
CAS 16503-22-3
Synonym N-Methyl-1H-imidazole-4-ethanamine dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature