missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylfurohydroxamic Acid [Chelating Reagent for HPLC], TCI America™

$249.03 - $747.09

Chemical Identifiers

CAS 109531-96-6
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00144939
InChI Key KCAHIEOWHDMBKM-UHFFFAOYSA-N
Synonym N-(2-Furoyl)-N-methylhydroxylamine, N-MFHA
PubChem CID 565703
IUPAC Name N-hydroxy-N-methylfuran-2-carboxamide
SMILES CN(C(=O)C1=CC=CO1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A50821G View Documents TCI America
A50821G
1g Each for $249.03
 
A50825G View Documents TCI America
A50825G
5g Each for $747.09
 
Specifications

Chemical Identifiers

109531-96-6
141.126
KCAHIEOWHDMBKM-UHFFFAOYSA-N
565703
CN(C(=O)C1=CC=CO1)O
C6H7NO3
MFCD00144939
N-(2-Furoyl)-N-methylhydroxylamine, N-MFHA
N-hydroxy-N-methylfuran-2-carboxamide

Specifications

White
1g
C6H7NO3
N-(2-Furoyl)-N-methylhydroxylamine, N-MFHA
CN(C(=O)C1=CC=CO1)O
141.126
141.13
≥99.0% (HPLC)
101°C
109531-96-6
MFCD00144939
KCAHIEOWHDMBKM-UHFFFAOYSA-N
N-hydroxy-N-methylfuran-2-carboxamide
565703
Crystal-Powder at 20°C
N-Methylfurohydroxamic Acid [Chelating Reagent for HPLC]
Safety and Handling

Safety and Handling

TSCA : Yes