missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylformanilide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 126810010

 View more versions of this product

Catalog No. AC126810010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Methylformanilide
93-61-8
100.0
Colorless to Yellow
243.0°C to 244.0°C
Authentic
Glass bottle
1.5600 to 1.5620
MFCD00003283
12,234
1.09
Solubility in water: immiscible.
CN(C=O)C1=CC=CC=C1
135.17
135.17
Liquid
99%
98.5
8.0°C to 13.0°C
1.0900g/mL
126°C
98.5% min. (GC)
C8H9NO
HCON(CH3)C6H5
1 kg
01,68
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
JIKUXBYRTXDNIY-UHFFFAOYSA-N
N-methyl-N-phenylformamide
66737
99%

Chemical Identifiers

93-61-8
JIKUXBYRTXDNIY-UHFFFAOYSA-N
66737
CN(C=O)C1=CC=CC=C1
MFCD00003283
n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
N-methyl-N-phenylformamide
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 202-262-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only