missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylformamide, 99+%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 414330050

 View more versions of this product

Catalog No. AC414330050


 

Specifications Description & Specifications

Specifications

N-Methylformamide
5g
123-39-7
100.0
Authentic
1.4310 to 1.4330 (20°C, 589nm)
≥99.0%
C2H5NO
59.06
04, 58
Clear colorless
99+%
99.0
1.0030g/mL
Glass bottle
Liquid
99+%
HCONHCH3
MFCD00003280
15, 6147

Chemical Identifiers

123-39-7
100.0
C2H5NO
59.06
N-Methylformamide, 99+%
99.0
99+%
HCONHCH3
MFCD00003280
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS P Statement Harmful in contact with skin. May damage the unborn child. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 204-624-6

RTECSNumber : LQ3000000

TSCA : TSCA