missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylethylenediamine, TCI America™

Manufacturer:  TCI America M0518100ML

 View more versions of this product

Catalog No. M0518100ML



Specifications

Specifications

N-Methylethylenediamine
Colorless
109-81-9
MFCD00008165
n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
CNCCN
74.127
74.13
≥97.0% (T)
116°C
100mL
C3H10N2
2734
KFIGICHILYTCJF-UHFFFAOYSA-N
N'-methylethane-1,2-diamine
8014
Clear Liquid at 20°C

Chemical Identifiers

109-81-9
74.127
KFIGICHILYTCJF-UHFFFAOYSA-N
8014
CNCCN
C3H10N2
MFCD00008165
n-methylethylenediamine, 1,2-ethanediamine, n-methyl, 2-aminoethylmethylamine, n-methyldiaminoethane, n-methylethanediamine, 2-methylamino ethylamine, n-methyl-1,2-ethanediamine, n-methylethylidenediamine, ethylenediamine, n-methyl, n-methylethane-1,2-diamine
N'-methylethane-1,2-diamine
Safety and Handling

Safety and Handling

RTECSNumber : KV5250500

TSCA : Yes

Documents