Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylethylenediamine, 98%

Chemical Identifiers

Chemical Name or Material N-Methylethylenediamine
CAS 109-81-9
Formula Weight 74.13
MDL Number MFCD00008165
UN Number UN2733
Synonym 2-Methylaminoethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3186604

 
alfa aesar™
3186604
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.13
Chemical Name or Material N-Methylethylenediamine
Refractive Index 1.4395
TSCA TSCA
Density 0.841g/mL
Boiling Point 114° to 116°C
Physical Form Liquid
Assay 98%
Quantity 2g
Flash Point 41° (106°F)
Odor Ammonia-like
MDL Number MFCD00008165
CAS 109-81-9
UN Number UN2733
Synonym 2-Methylaminoethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.