missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyldiphenylamine, TCI America™

Manufacturer:  TCI America M021125ML

 View more versions of this product

Catalog No. M021125ML



Specifications

Specifications

N-Methyldiphenylamine
Yellow
552-82-9
MFCD00041900
DYFFAVRFJWYYQO-UHFFFAOYSA-N
N-methyl-N-phenylaniline
11098
Clear Liquid at 20°C
295°C
25mL
C13H13N
n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
CN(C1=CC=CC=C1)C2=CC=CC=C2
183.254
183.25
≥98.0% (GC)

Chemical Identifiers

552-82-9
183.254
DYFFAVRFJWYYQO-UHFFFAOYSA-N
11098
CN(C1=CC=CC=C1)C2=CC=CC=C2
C13H13N
MFCD00041900
n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
N-methyl-N-phenylaniline
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0364

TSCA : Yes

Documents