missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyldiphenylamine, TCI America™

$67.22 - $172.64

Chemical Identifiers

CAS 552-82-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00041900
InChI Key DYFFAVRFJWYYQO-UHFFFAOYSA-N
Synonym n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
PubChem CID 11098
IUPAC Name N-methyl-N-phenylaniline
SMILES CN(C1=CC=CC=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M02115ML View Documents TCI America
M02115ML
5mL Each for $67.22
 
M021125ML View Documents TCI America
M021125ML
25mL Each for $172.64
 
Specifications

Chemical Identifiers

552-82-9
183.254
DYFFAVRFJWYYQO-UHFFFAOYSA-N
11098
CN(C1=CC=CC=C1)C2=CC=CC=C2
C13H13N
MFCD00041900
n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
N-methyl-N-phenylaniline

Specifications

295°C
5mL
C13H13N
n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
CN(C1=CC=CC=C1)C2=CC=CC=C2
183.254
183.25
≥98.0% (GC)
Yellow
552-82-9
MFCD00041900
DYFFAVRFJWYYQO-UHFFFAOYSA-N
N-methyl-N-phenylaniline
11098
Clear Liquid at 20°C
N-Methyldiphenylamine
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0364

TSCA : Yes