Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyldiacetamide, 90+%

$427.92 - $427.92

Chemical Identifiers

Chemical Name or Material N-Methyldiacetamide
CAS 1113-68-4
Molecular Formula (CH3CO)2NCH3
Formula Weight 115.13
MDL Number MFCD00014968
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2029114

  SDS
alfa aesar™
B2029114
25g Each for $427.92
Description & Specifications

Specifications

Formula Weight 115.13
TSCA TSCA
CAS 1113-68-4
Quantity 25g
Assay ≥90%
Molecular Formula (CH3CO)2NCH3
MDL Number MFCD00014968
Chemical Name or Material N-Methyldiacetamide
Boiling Point 192° to 194°C

Applications
It can be used to produce N-Methyl-N-[1-(trimethylsiloxy)vinyl]acetamid at the ambient temperature.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.