Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyldi-n-propylamine, 98%

Chemical Identifiers

Chemical Name or Material N-Methyldi-n-propylamine
CAS 3405-42-3
Linear Formula (CH3CH2CH2)2NCH3
Formula Weight 115.22
MDL Number MFCD00048683
UN Number UN2924
Beilstein 1731704
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6507706

  SDS
alfa aesar™
H6507706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6507714

  SDS
alfa aesar™
H6507714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.22
Boiling Point 113° to 117°C
Chemical Name or Material N-Methyldi-n-propylamine
Refractive Index 1.4060
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless
Physical Form Liquid
Assay 98%
Linear Formula (CH3CH2CH2)2NCH3
MDL Number MFCD00048683
CAS 3405-42-3
UN Number UN2924
Beilstein 1731704
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Density 0.743g/mL
Quantity 5g

Applications
N-Methyldi-n-propylamine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot water.

Notes
Do not store together with acids. Incompatible with acids and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.