missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylcyclohexylamine 99.0+%, TCI America™

Manufacturer:  TCI America M0202500G

 View more versions of this product

Catalog No. M0202500G



Specifications

Specifications

N-Methylcyclohexylamine
Colorless
100-60-7
MFCD00003832
n-methylcyclohexylamine, methylcyclohexylamine, n-cyclohexylmethylamine, cyclohexanamine, n-methyl, n-methyl-n-cyclohexylamine, methylamino cyclohexane, cyclohexylamine, n-methyl, cyclohexyl methyl amine, n-cyclohexyl-n-methylamine, unii-yh8pf4c1ow
CNC1CCCCC1
113.204
CHEBI:59018
Clear Liquid at 20°C
148°C
500g
C7H15N
2734
XTUVJUMINZSXGF-UHFFFAOYSA-N
N-methylcyclohexanamine
7514
113.20
≥99.0% (T)

Chemical Identifiers

100-60-7
113.204
XTUVJUMINZSXGF-UHFFFAOYSA-N
7514
N-methylcyclohexanamine
C7H15N
MFCD00003832
n-methylcyclohexylamine, methylcyclohexylamine, n-cyclohexylmethylamine, cyclohexanamine, n-methyl, n-methyl-n-cyclohexylamine, methylamino cyclohexane, cyclohexylamine, n-methyl, cyclohexyl methyl amine, n-cyclohexyl-n-methylamine, unii-yh8pf4c1ow
CHEBI:59018
CNC1CCCCC1
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4003

RTECSNumber : GX1529000

TSCA : Yes

Documents