Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylcyclohexylamine, 98%

Chemical Identifiers

Chemical Name or Material N-Methylcyclohexylamine
CAS 100-60-7
Molecular Formula C7H15N
Formula Weight 113.2
MDL Number MFCD00003832
UN Number UN2734
Beilstein 1523664
Synonym N-Cyclohexylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL13872AE

  SDS
alfa aesar™
L13872AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.2
Chemical Name or Material N-Methylcyclohexylamine
Assay 98%
MDL Number MFCD00003832
CAS 100-60-7
UN Number UN2734
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Melting Point −8°C
Boiling Point 149°C
Quantity 100mL
Flash Point 35°C (95°F)
Molecular Formula C7H15N
Beilstein 1523664
Synonym N-Cyclohexylmethylamine
Health Hazard 2 GHS H Statement
H301-H314-H318-H226-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
RTECS Number GX1529000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H226-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.