missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylcarbazole, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441050250

 View more versions of this product

Catalog No. AC441050250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Methylcarbazole
163.0°C to 167.0°C (0.4 Torr)
86.0°C to 89.0°C
1484-12-4
100.0
C13H11N
SDFLTYHTFPTIGX-UHFFFAOYSA-N
9-methylcarbazole
15142
Crystalline Powder
Authentic
White
25g
98.5
98.5% min. (HPLC)
9-methyl-9h-carbazole, 9-methylcarbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, ccris 6846, n-methyl carbazole, n-methylcarbazole, n-methyldibenzopyrrole, pubchem17421, unii-76sop090pg
CN1C2=CC=CC=C2C3=CC=CC=C31
181.24
181.24
99%

Chemical Identifiers

1484-12-4
181.24
9-methyl-9h-carbazole, 9-methylcarbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, ccris 6846, n-methyl carbazole, n-methylcarbazole, n-methyldibenzopyrrole, pubchem17421, unii-76sop090pg
9-methylcarbazole
C13H11N
SDFLTYHTFPTIGX-UHFFFAOYSA-N
15142
CN1C2=CC=CC=C2C3=CC=CC=C31
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-054-5

SDS Documents