missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylbenzenesulfonamide, TCI America™

$151.25 - $1,126.00

Chemical Identifiers

CAS 5183-78-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00025023
InChI Key SVDVKEBISAOWJT-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-methyl, n-methylbenzenesulphonamide, n-phenylsulfonyl methanamine, methyl phenylsulfonyl amine, n-methylphenylsulfonamide, acmc-1avn9, n-methylbenzene sulfonamide, n-methyl benzenesulphonamide, benzenesulfonamide,n-methyl, benzenesulfonyl-n-methylamide
PubChem CID 78858
IUPAC Name N-methylbenzenesulfonamide
SMILES CNS(=O)(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M015725G View Documents TCI America
M015725G
25g Each for $151.25
 
M0157500G View Documents TCI America
M0157500G
500g Each for $1,126.00
 
Specifications

Chemical Identifiers

5183-78-8
171.214
SVDVKEBISAOWJT-UHFFFAOYSA-N
78858
CNS(=O)(=O)C1=CC=CC=C1
C7H9NO2S
MFCD00025023
benzenesulfonamide, n-methyl, n-methylbenzenesulphonamide, n-phenylsulfonyl methanamine, methyl phenylsulfonyl amine, n-methylphenylsulfonamide, acmc-1avn9, n-methylbenzene sulfonamide, n-methyl benzenesulphonamide, benzenesulfonamide,n-methyl, benzenesulfonyl-n-methylamide
N-methylbenzenesulfonamide

Specifications

White-Yellow
5183-78-8
MFCD00025023
SVDVKEBISAOWJT-UHFFFAOYSA-N
N-methylbenzenesulfonamide
78858
Crystal at 20°C
N-Methylbenzenesulfonamide
25g
C7H9NO2S
benzenesulfonamide, n-methyl, n-methylbenzenesulphonamide, n-phenylsulfonyl methanamine, methyl phenylsulfonyl amine, n-methylphenylsulfonamide, acmc-1avn9, n-methylbenzene sulfonamide, n-methyl benzenesulphonamide, benzenesulfonamide,n-methyl, benzenesulfonyl-n-methylamide
CNS(=O)(=O)C1=CC=CC=C1
171.214
171.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1930

TSCA : Yes