Please login to your online account to display your discounted pricing

N-Methylanilinium trifluoroacetate, 99%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  198421000

 View more versions of this product

Catalog No. AC198421000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Needles
Assay ≥98.5%
Formula Weight 221.18
Color White to very slightly grayish
Quantity 100g
EINECS Number 249-927-4
Melting Point 64° to 67°C
Fieser 08,331
CAS Max % 100.0
Chemical Name or Material N-Methylanilinium trifluoroacetate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H10N·C2F3O2
Linear Formula {C6H5NH2(CH3)}O2CCF3
MDL Number MFCD00043278
CAS 29885-95-8
Synonym TAMA
CAS Min % 98.5
Health Hazard 1 HYGROSCOPIC
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-Methylanilinium trifluoroacetate
Name Note 99%
CAS 29885-95-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10N·C2F3O2
Linear Formula {C6H5NH2(CH3)}O2CCF3
Formula Weight 221.18
MDL Number MFCD00043278
Synonym TAMA
Accessories

HYGROSCOPIC

SDS & Safety Documents