Please login to your online account to display your discounted pricing

N-Methylanilinium trifluoroacetate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Methylanilinium trifluoroacetate
Name Note 99%
CAS 29885-95-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10N·C2F3O2
Linear Formula {C6H5NH2(CH3)}O2CCF3
Formula Weight 221.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC198421000

  SDS
thermo fisher scientific
198421000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Needles
Assay ≥98.5%
Color White to very slightly grayish
Formula Weight 221.18
Quantity 100g
EINECS Number 249-927-4
Melting Point 64° to 67°C
Fieser 08,331
CAS Max % 100.0
Synonym TAMA
CAS Min % 98.5
Health Hazard 1 HYGROSCOPIC
Infrared Spectrum Authentic
Assay Percent Range 99%
CAS 29885-95-8
Chemical Name or Material N-Methylanilinium trifluoroacetate
Linear Formula {C6H5NH2(CH3)}O2CCF3
Name Note 99%
Molecular Formula C7H10N·C2F3O2
MDL Number MFCD00043278
Safety and Handling

HYGROSCOPIC