Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylanilinium trifluoroacetate, 99%

Chemical Identifiers

Chemical Name or Material N-Methylanilinium trifluoroacetate
CAS 29885-95-8
Molecular Formula C7H10N·CF3CO2
Formula Weight 221.18
MDL Number MFCD00043278
UN Number UN3077
Beilstein 3638133
Synonym TAMA; Trifluoroacetic acid N-methylanilinium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1778114

  SDS
alfa aesar™
A1778114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1778122

  SDS
alfa aesar™
A1778122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1778136

  SDS
alfa aesar™
A1778136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.18
Assay 99%
Molecular Formula C7H10N·CF3CO2
Quantity 25g
UN Number UN3077
Health Hazard 1 Danger
Synonym TAMA; Trifluoroacetic acid N-methylanilinium salt
Chemical Name or Material N-Methylanilinium trifluoroacetate
MDL Number MFCD00043278
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 29885-95-8
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Melting Point 64° to 68°C
Beilstein 3638133
Health Hazard 2 GHS H Statement
H301-H311-H373-H332
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H373-H332
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.