Learn More
N-Methylanilinium trifluoroacetate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1778114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Methylanilinium trifluoroacetate | |
64°C to 68°C | |
MFCD00043278 | |
UN3077 | |
Hygroscopic | |
221.18 |
29885-95-8 | |
C7H10N·CF3CO2 | |
25 g | |
3638133 | |
TAMA; Trifluoroacetic acid N-methylanilinium salt | |
99% |
Chemical Identifiers
29885-95-8 | |
C7H10N·CF3CO2 | |
MFCD00043278 | |
TAMA; Trifluoroacetic acid N-methylanilinium salt |
Safety and Handling
GHS H Statement
H301-H311-H373-H332
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 249-927-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only