Learn More
N-Methylaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1387322
Description
N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
Specifications
N-Methylaniline | |
-57°C | |
194°C to 197°C | |
C7H9N | |
MFCD00008283 | |
UN2294 | |
Air Sensitive | |
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane | |
AFBPFSWMIHJQDM-UHFFFAOYSA-N | |
N-methylaniline | |
7515 | |
107.16 |
100-61-8 | |
0.989 | |
86°C (186°F) | |
1.571 | |
100 g | |
741982 | |
14,6019 | |
Immiscible with water. | |
CNC1=CC=CC=C1 | |
107.16 | |
CHEBI:15733 | |
98% |
Chemical Identifiers
100-61-8 | |
107.16 | |
AFBPFSWMIHJQDM-UHFFFAOYSA-N | |
7515 | |
N-methylaniline |
C7H9N | |
MFCD00008283 | |
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane | |
CHEBI:15733 | |
CNC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-METHYLANILINE
EINECSNumber : 202-870-9
RTECSNumber : BY4550000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only