missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylaniline, 98%

Manufacturer:  Alfa Aesar™ L1387336

 View more versions of this product

Catalog No. AAL1387336



Description

Description

Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
Specifications

Specifications

N-Methylaniline
194°C to 197°C
500g
C7H9N
UN2294
14,6019
AFBPFSWMIHJQDM-UHFFFAOYSA-N
N-methylaniline
7515
107.16
86°C (186°F)
-57°C
100-61-8
MFCD00008283
741982
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
CNC1=CC=CC=C1
107.156
CHEBI:15733
98%

Chemical Identifiers

100-61-8
107.156
AFBPFSWMIHJQDM-UHFFFAOYSA-N
7515
N-methylaniline
C7H9N
MFCD00008283
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
CHEBI:15733
CNC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-METHYLANILINE

RTECSNumber : BY4550000

TSCA : TSCA

Documents