Please login to your online account to display your discounted pricing

N-Methylacetamide, 99+%, ACROS Organics™

$154.44 - $154.44

Chemical Identifiers

Chemical Name or Material N-Methylacetamide
Name Note 99+%
CAS 79-16-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C3H7NO
Linear Formula CH3CONHCH3
Formula Weight 73.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126140010

  SDS
thermo fisher scientific
126140010
Glass bottle 1kg Each for $154.44
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid After Melting
Formula Weight 73.1
Quantity 1kg
Color Clear colorless to yellow
Beilstein 04, 58
EINECS Number 201-182-6
TSCA TSCA
RTECS Number AC5960000
Density 0.946g/mL
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May damage the unborn child.
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
Linear Formula CH3CONHCH3
MDL Number MFCD00008683
Name Note 99+%
Chemical Name or Material N-Methylacetamide
Molecular Formula C3H7NO
Assay Percent Range 99+%
CAS 79-16-3
Safety and Handling

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child.