Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylacetamide, 99%

$41.58 - $965.84

Chemical Identifiers

Chemical Name or Material N-Methylacetamide
CAS 79-16-3
Molecular Formula CH3C(O)NHCH3
Formula Weight 73.1
MDL Number MFCD00008683
Beilstein 1071255
Synonym N-Acetylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA179280C

  SDS
alfa aesar™
A179280C
10000g Each for $965.84

AAA179280E

  SDS
alfa aesar™
A179280E
2500g Each for $265.19

AAA1792830

  SDS
alfa aesar™
A1792830
250g Each for $41.58

AAA1792836

  SDS
alfa aesar™
A1792836
500g Each for $69.76
Description & Specifications

Specifications

Formula Weight 73.1
Flash Point 116°C (240°F)
Boiling Point 204° to 206°C
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 79-16-3
Health Hazard 1 Danger
Beilstein 1071255
Odor Odorless
Molecular Formula CH3C(O)NHCH3
Assay 99%
RTECS Number AC5960000
Quantity 10000g
TSCA TSCA
Synonym N-Acetylmethylamine
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Melting Point 26° to 28°C
MDL Number MFCD00008683
Chemical Name or Material N-Methylacetamide

Applications
N-Methylacetamide is used as a chemical intermediate in the production of life science, agrochemicals, electronic materials and construction materials. It is also used as a solvent, in electrochemistry.

Solubility
Soluble in water (1X10+6 mg/L @ 20 deg C), chloroform. Soluble in alcohol, ether, acetone, benzene.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.