missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-(tetrahydropyran-4-ylmethyl)amine, 97%, Maybridge

Manufacturer:  Thermo Scientific CC29946DA

 View more versions of this product

Catalog No. CC29946DA



Specifications

Specifications

N-methyl-(tetrahydropyran-4-ylmethyl)amine
1g
96.5
97%
MFCD06739009
WMBCUXKYKVTJRF-UHFFFAOYSA-N
N-methyl-1-(oxan-4-yl)methanamine
22225668
97%
Amber Glass Bottle
439081-52-4
100.0
C7H15NO
methyl oxan-4-ylmethyl amine, methyl-tetrahydropyran-4-ylmethyl amine, n-methyl-1-tetrahydro-2h-pyran-4-yl methanamine, methyl-tetrahydro-pyran-4-ylmethyl-amine, n-methyl-tetrahydropyran-4-ylmethyl amine, 4-methylamino methyl tetrahydropyran, 2h-pyran-4-methanamine, tetrahydro-n-methyl, n-methyl-1-tetrahydro-2h-pyran-4-yl methylamine, n-methyl tetrahydro-2h-pyran-4-yl methanamine, 4-methylamino methyl tetrahydro-2h-pyran
CNCC1CCOCC1
129.203
129.2

Chemical Identifiers

439081-52-4
129.203
WMBCUXKYKVTJRF-UHFFFAOYSA-N
22225668
CNCC1CCOCC1
C7H15NO
MFCD06739009
methyl oxan-4-ylmethyl amine, methyl-tetrahydropyran-4-ylmethyl amine, n-methyl-1-tetrahydro-2h-pyran-4-yl methanamine, methyl-tetrahydro-pyran-4-ylmethyl-amine, n-methyl-tetrahydropyran-4-ylmethyl amine, 4-methylamino methyl tetrahydropyran, 2h-pyran-4-methanamine, tetrahydro-n-methyl, n-methyl-1-tetrahydro-2h-pyran-4-yl methylamine, n-methyl tetrahydro-2h-pyran-4-yl methanamine, 4-methylamino methyl tetrahydro-2h-pyran
N-methyl-1-(oxan-4-yl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents