missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-tert-butylamine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC368810250

 View more versions of this product

Catalog No. AC368810250



Specifications

Specifications

N-Methyl-tert-butylamine
−26°C
1.3950 to 1.3970
67.0°C to 69.0°C
97%
96.0
96% (GC)
(CH3)3CNHCH3
04,IV,659
ZQGJEUVBUVKZKS-UHFFFAOYSA-N
N,2-dimethylpropan-2-amine
84552
97%
0.7270g/mL
Glass Bottle
0.727
25mL
14610-37-8
100.0
C5H13N
MFCD00042853
2-propanamine, n,2-dimethyl
CC(C)(C)NC
87.16
87.16

Chemical Identifiers

14610-37-8
87.16
ZQGJEUVBUVKZKS-UHFFFAOYSA-N
84552
CC(C)(C)NC
C5H13N
MFCD00042853
2-propanamine, n,2-dimethyl
N,2-dimethylpropan-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 238-646-2

SDS Documents