missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl Paroxetine, TCI America™

$169.59 - $568.34

Chemical Identifiers

CAS 110429-36-2
Molecular Formula C20H22FNO3
Molecular Weight (g/mol) 343.398
MDL Number MFCD03788781
InChI Key MOJZPKOBKCXNKG-YJBOKZPZSA-N
PubChem CID 10947895
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)-1-methylpiperidine
SMILES CN1CCC(C(C1)COC2=CC3=C(C=C2)OCO3)C4=CC=C(C=C4)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2645200MG View Documents TCI America
M2645200MG
200mg Each for $169.59
 
M26451G View Documents TCI America
M26451G
1g Each for $568.34
 
Specifications

Chemical Identifiers

110429-36-2
343.398
MOJZPKOBKCXNKG-YJBOKZPZSA-N
(3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)-1-methylpiperidine
C20H22FNO3
MFCD03788781
10947895
CN1CCC(C(C1)COC2=CC3=C(C=C2)OCO3)C4=CC=C(C=C4)F

Specifications

White
200mg
C20H22FNO3
MOJZPKOBKCXNKG-YJBOKZPZSA-N
(3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)-1-methylpiperidine
10947895
Crystal-Powder at 20°C
N-Methyl Paroxetine
111°C
110429-36-2
MFCD03788781
CN1CCC(C(C1)COC2=CC3=C(C=C2)OCO3)C4=CC=C(C=C4)F
343.398
343.40
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer