missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-p-toluidine, TCI America™

$62.64 - $369.73

Chemical Identifiers

CAS 623-08-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00025628
InChI Key QCIFLGSATTWUQJ-UHFFFAOYSA-N
Synonym N,4-Dimethylaniline, 4-(Methylamino)toluene
PubChem CID 12165
IUPAC Name N,4-dimethylaniline
SMILES CC1=CC=C(C=C1)NC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M131525ML View Documents TCI America
M131525ML
25mL Each for $62.64
 
M1315250ML View Documents TCI America
M1315250ML
250mL Each for $369.73
 
Specifications

Chemical Identifiers

623-08-5
121.183
QCIFLGSATTWUQJ-UHFFFAOYSA-N
12165
CC1=CC=C(C=C1)NC
C8H11N
MFCD00025628
N,4-Dimethylaniline, 4-(Methylamino)toluene
N,4-dimethylaniline

Specifications

212°C
25mL
C8H11N
2810
QCIFLGSATTWUQJ-UHFFFAOYSA-N
N,4-dimethylaniline
12165
Clear Liquid at 20°C
N-Methyl-p-toluidine
Yellow
623-08-5
MFCD00025628
N,4-Dimethylaniline, 4-(Methylamino)toluene
CC1=CC=C(C=C1)NC
121.183
121.18
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0129&(3)-0187

TSCA : Yes