missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-p-toluidine 97.0+%, TCI America™
Supplier: TCI America M1315250ML
Specifications
N-Methyl-p-toluidine | |
Yellow | |
C8H11N | |
250 mL | |
N,4-Dimethylaniline, 4-(Methylamino)toluene | |
CNC1=CC=C(C)C=C1 | |
121.18 | |
121.18 | |
Liquid |
623-08-5 | |
212°C | |
MFCD00025628 | |
2810 | |
QCIFLGSATTWUQJ-UHFFFAOYSA-N | |
N,4-dimethylaniline | |
12165 | |
≥97.0% (GC) |
Chemical Identifiers
623-08-5 | |
121.18 | |
QCIFLGSATTWUQJ-UHFFFAOYSA-N | |
12165 | |
CNC1=CC=C(C)C=C1 |
C8H11N | |
MFCD00025628 | |
N,4-Dimethylaniline, 4-(Methylamino)toluene | |
N,4-dimethylaniline |
Safety and Handling
EINECSNumber : (3)-0129&(3)-0187
TSCA : Yes