missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-p-toluenesulfonamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC164845000

 View more versions of this product

Catalog No. AC164845000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

N-Methyl-p-toluenesulfonamide
76° to 80°C
640-61-9
100.0
Glass bottle
≥97.5%
C8H11NO2S
185.24
11, 105
White to light yellow
500g
97.5
Authentic
Crystalline Solid
98%
CH3C6H4SO2NHCH3
MFCD00008285

Chemical Identifiers

640-61-9
100.0
C8H11NO2S
185.24
N-Methyl-p-toluenesulfonamide
97.5
98%
CH3C6H4SO2NHCH3
MFCD00008285
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.