Please login to your online account to display your discounted pricing

N-Methyl-p-toluenesulfonamide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Methyl-p-toluenesulfonamide
CAS 640-61-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H11NO2S
Linear Formula CH3C6H4SO2NHCH3
Formula Weight 185.24
MDL Number MFCD00008285
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC164845000

  SDS
thermo fisher scientific
AC164845000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC164840500

  SDS
thermo fisher scientific
164840500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC164840100

  SDS
fisher scientific acros
164840100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Solid
Formula Weight 185.24
Quantity 500g
Color White to light yellow
Melting Point 76° to 80°C
Beilstein 11, 105
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula CH3C6H4SO2NHCH3
MDL Number MFCD00008285
CAS Min % 97.5
Chemical Name or Material N-Methyl-p-toluenesulfonamide
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C8H11NO2S
CAS 640-61-9
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.