missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-trimethylsilylacetamide 90.0+%, TCI America™

$72.61 - $141.98

Chemical Identifiers

CAS 7449-74-3
Molecular Formula C6H15NOSi
Molecular Weight (g/mol) 145.277
MDL Number MFCD00040758
InChI Key QHUOBLDKFGCVCG-UHFFFAOYSA-N
Synonym acetamide, n-methyl-n-trimethylsilyl, acmc-1bei6, n-methyl-n-tms-acetamide, n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc, n-methylacetamide, tms derivative, n-trimethylsilyl-n-methylacetamide, qhuobldkfgcvcg-uhfffaoysa
PubChem CID 81953
IUPAC Name N-methyl-N-trimethylsilylacetamide
SMILES CC(=O)N(C)[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M053610G View Documents Tci America
M053610G
10g Each for $72.61
 
M053625G View Documents Tci America
M053625G
25g Each for $141.98
 
Specifications

Chemical Identifiers

7449-74-3
145.277
QHUOBLDKFGCVCG-UHFFFAOYSA-N
81953
CC(=O)N(C)[Si](C)(C)C
C6H15NOSi
MFCD00040758
acetamide, n-methyl-n-trimethylsilyl, acmc-1bei6, n-methyl-n-tms-acetamide, n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc, n-methylacetamide, tms derivative, n-trimethylsilyl-n-methylacetamide, qhuobldkfgcvcg-uhfffaoysa
N-methyl-N-trimethylsilylacetamide

Specifications

154°C
10g
C6H15NOSi
2924
QHUOBLDKFGCVCG-UHFFFAOYSA-N
N-methyl-N-trimethylsilylacetamide
81953
Clear Liquid at 20°C
N-Methyl-N-trimethylsilylacetamide [Trimethylsilylating Agent]
Yellow
7449-74-3
MFCD00040758
acetamide, n-methyl-n-trimethylsilyl, acmc-1bei6, n-methyl-n-tms-acetamide, n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc, n-methylacetamide, tms derivative, n-trimethylsilyl-n-methylacetamide, qhuobldkfgcvcg-uhfffaoysa
CC(=O)N(C)[Si](C)(C)C
145.277
145.28
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator