Please login to your online account to display your discounted pricing

N-Methyl-N-(trimethylsilyl)acetamide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Methyl-N-(trimethylsilyl)acetamide
Name Note 98%
CAS 7449-74-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H15NOSi
Linear Formula CH3CON(CH3)Si(CH3)3
Formula Weight 145.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215950010

  SDS
thermo fisher scientific
AC215950010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 145.28
Quantity 1g
Color Clear light yellow
EINECS Number 231-217-0
TSCA TSCA
Density 0.9g/mL
Beilstein 04, IV, 4011
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material N-Methyl-N-(trimethylsilyl)acetamide
Infrared Spectrum Authentic
Name Note 98%
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C6H15NOSi
Linear Formula CH3CON(CH3)Si(CH3)3
MDL Number MFCD00040758
CAS 7449-74-3
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Silylation reagent

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.