Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-n-propylamine

Chemical Identifiers

Chemical Name or Material N-Methyl-n-propylamine
CAS 627-35-0
Formula Weight 73.14g/mol
MDL Number MFCD00009361
UN Number UN2733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1562503

 
alfa aesar™
15625-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.713g/mL
Boiling Point 62° to 64°C
Quantity 1g
Flash Point −1° (30°F)
Odor Amine-like
Chemical Name or Material N-Methyl-n-propylamine
CAS 627-35-0
Physical Form Liquid
Formula Weight 73.14g/mol
MDL Number MFCD00009361
UN Number UN2733
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.