missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-propargylbenzylamine, TCI America™

$304.03 - $909.05

Chemical Identifiers

CAS 555-57-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00008576
InChI Key DPWPWRLQFGFJFI-UHFFFAOYSA-N
Synonym pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
PubChem CID 4688
ChEBI CHEBI:7930
IUPAC Name N-benzyl-N-methylprop-2-yn-1-amine
SMILES CN(CC#C)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26181G View Documents TCI America
M26181G
1g Each for $304.03
 
M26185G View Documents TCI America
M26185G
5g Each for $909.05
 
Specifications

Chemical Identifiers

555-57-7
159.232
DPWPWRLQFGFJFI-UHFFFAOYSA-N
4688
N-benzyl-N-methylprop-2-yn-1-amine
C11H13N
MFCD00008576
pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
CHEBI:7930
CN(CC#C)CC1=CC=CC=C1

Specifications

90°C
1g
C11H13N
2810
DPWPWRLQFGFJFI-UHFFFAOYSA-N
N-benzyl-N-methylprop-2-yn-1-amine
4688
159.23
≥98.0% (GC,T)
Yellow
555-57-7
MFCD00008576
pargyline, pargylamine, n-methyl-n-propargylbenzylamine, paragyline, eudatin, supirdyl, eutonyl, pargylin, n-methyl-n-2-propynylbenzylamine, benzyl-methyl-2-propinylamin
CN(CC#C)CC1=CC=CC=C1
159.232
CHEBI:7930
Clear Liquid at 20°C
N-Methyl-N-propargylbenzylamine
Safety and Handling

Safety and Handling

RTECSNumber : DP6475000

TSCA : No