Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methyl-N-phenylthiourea, 97%

Chemical Identifiers

Chemical Name or Material N-Methyl-N-phenylthiourea
CAS 4104-75-0
Molecular Formula C8H10N2S
Formula Weight 166.24
MDL Number MFCD00086629
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2082409

  SDS
alfa aesar™
B2082409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2082418

  SDS
alfa aesar™
B2082418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.24
Odor Odorless
Chemical Name or Material N-Methyl-N-phenylthiourea
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 100° to 104°C
Quantity 10g
CAS 4104-75-0
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
MDL Number MFCD00086629
Molecular Formula C8H10N2S
UN Number UN2811
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.

Applications
It is used to produce S-Ethyl-N-methyl-N-phenyl-isothiourea; hydriodide by heating. In the presence of solvent acetone.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.