missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-nitrosourethane, TCI America™

$102.36 - $279.59

Chemical Identifiers

CAS 615-53-2
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00013888
InChI Key CAUBWLYZCDDYEF-UHFFFAOYSA-N
Synonym N-Ethoxycarbonyl-N-methylnitrosamine, N-Nitroso-N-methylurethane, Ethyl N-Methyl-N-nitrosocarbamate
PubChem CID 12001
ChEBI CHEBI:82373
IUPAC Name ethyl N-methyl-N-nitrosocarbamate
SMILES CCOC(=O)N(C)N=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N02655G View Documents TCI America
N02655G
5g Each for $102.36
 
N026525G View Documents TCI America
N026525G
25g Each for $279.59
 
Specifications

Chemical Identifiers

615-53-2
132.119
CAUBWLYZCDDYEF-UHFFFAOYSA-N
12001
ethyl N-methyl-N-nitrosocarbamate
C4H8N2O3
MFCD00013888
N-Ethoxycarbonyl-N-methylnitrosamine, N-Nitroso-N-methylurethane, Ethyl N-Methyl-N-nitrosocarbamate
CHEBI:82373
CCOC(=O)N(C)N=O

Specifications

66°C
5g
C4H8N2O3
2810
CAUBWLYZCDDYEF-UHFFFAOYSA-N
ethyl N-methyl-N-nitrosocarbamate
12001
132.12
≥95.0% (GC)
Undesignated
615-53-2
MFCD00013888
N-Ethoxycarbonyl-N-methylnitrosamine, N-Nitroso-N-methylurethane, Ethyl N-Methyl-N-nitrosocarbamate
CCOC(=O)N(C)N=O
132.119
CHEBI:82373
Clear Liquid at 20°C
N-Methyl-N-nitrosourethane
Safety and Handling

Safety and Handling

RTECSNumber : FC6300000

TSCA : Yes

Recommended Storage : Refrigerator