missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-N-[(6-morpholin-4-ylpyridin-2-yl)methyl]amine 90+%, Thermo Scientific™
Chemical Identifiers
CAS |
886851-30-5 |
Molecular Formula |
C11H17N3O |
Molecular Weight (g/mol) |
207.28 |
MDL Number |
MFCD08435863 |
InChI Key |
CULRQHZBTPUFEB-UHFFFAOYSA-N |
Synonym |
n-methyl-n-6-morpholin-4-ylpyridin-2-yl methyl amine, methyl 6-morpholin-4-yl pyridin-2-yl methyl amine, methyl 6-morpholin-4-yl 2-pyridyl methyl amine, 2-pyridinemethanamine,n-methyl-6-4-morpholinyl, n-methyl-1-6-morpholin-4-yl pyridin-2-yl methanamine |
PubChem CID |
18525740 |
SMILES |
CNCC1=NC(=CC=C1)N1CCOCC1 |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
886851-30-5 |
|
207.28 |
|
CULRQHZBTPUFEB-UHFFFAOYSA-N |
|
18525740 |
|
C11H17N3O |
|
MFCD08435863 |
|
n-methyl-n-6-morpholin-4-ylpyridin-2-yl methyl amine, methyl 6-morpholin-4-yl pyridin-2-yl methyl amine, methyl 6-morpholin-4-yl 2-pyridyl methyl amine, 2-pyridinemethanamine,n-methyl-6-4-morpholinyl, n-methyl-1-6-morpholin-4-yl pyridin-2-yl methanamine |
|
CNCC1=NC(=CC=C1)N1CCOCC1 |
Specifications
|
886851-30-5 |
|
Amber Glass Bottle |
|
MFCD08435863 |
|
n-methyl-n-6-morpholin-4-ylpyridin-2-yl methyl amine, methyl 6-morpholin-4-yl pyridin-2-yl methyl amine, methyl 6-morpholin-4-yl 2-pyridyl methyl amine, 2-pyridinemethanamine,n-methyl-6-4-morpholinyl, n-methyl-1-6-morpholin-4-yl pyridin-2-yl methanamine |
|
CNCC1=NC(=CC=C1)N1CCOCC1 |
|
207.28 |
|
207.28 |
|
N-Methyl-N-[(6-morpholin-4-ylpyridin-2-yl)methyl]amine |
|
90% |
|
C11H17N3O |
|
250 mg |
|
CULRQHZBTPUFEB-UHFFFAOYSA-N |
|
N-methyl-1-(6-morpholin-4-ylpyridin-2-yl)methanamine |
|
18525740 |
|
90% |
SDS
missing translation for 'documents'