Please login to your online account to display your discounted pricing

N-Methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material N-methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine
CAS 869901-18-8
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C13H15NO
Formula Weight 201.27
MDL Number MFCD08690282
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

CC49746CB

  SDS
fisher scientific company
CC49746CB
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC49746DA

  SDS
fisher scientific company
CC49746DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 90%
Formula Weight 201.27
Quantity 250mg
CAS Max % 100.0
Chemical Name or Material N-methyl-N-[(5-methyl-2-phenyl-3-furyl)methyl]amine
Molecular Formula C13H15NO
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 90%
MDL Number MFCD08690282
CAS 869901-18-8
CAS Min % 89.5
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.