missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC35446DA
Specifications
N-Methyl-N-(4-pyridin-4-ylbenzyl)amine | |
90% | |
C13H14N2 | |
n-methyl-n-4-pyridin-4-ylbenzyl amine, n-methyl-4-pyridin-4-yl benzylamine, methyl 4-pyridin-4-yl phenyl methyl amine, n-methyl-1-4-pyridin-4-yl phenyl methanamine, n-methyl-1-4-pyridin-4-ylphenyl methanamine, methyl 4-4-pyridyl phenyl methyl amine, benzenemethanamine,n-methyl-4-4-pyridinyl | |
CNCC1=CC=C(C=C1)C2=CC=NC=C2 | |
198.269 | |
198.27 |
852180-64-4 | |
Amber Glass Bottle | |
1g | |
MDKJAMMSIZLRBH-UHFFFAOYSA-N | |
N-methyl-1-(4-pyridin-4-ylphenyl)methanamine | |
7060572 | |
90% |