Please login to your online account to display your discounted pricing

N-Methyl-N-[4-(piperidin-1-ylmethyl)benzyl]amine dihydrochloride, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material N-methyl-N-[4-(piperidin-1-ylmethyl)benzyl]amine dihydrochloride
CAS 859833-23-1
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C14H22N2·2ClH
Formula Weight 291.26
MDL Number MFCD07772864
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC45596DA

  SDS
fisher scientific company
CC45596DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.26
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C14H22N2·2ClH
MDL Number MFCD07772864
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Packaging Amber Glass Bottle
Chemical Name or Material N-methyl-N-[4-(piperidin-1-ylmethyl)benzyl]amine dihydrochloride
CAS 859833-23-1
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.