missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-[(4-methyl-4H-thieno[3,2-b]pyrrol-5-yl)methyl]amine hydrochloride, 95%, Maybridge

Chemical Identifiers

CAS 335033-68-6
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H12N2S·ClH
Formula Weight 216.73
MDL Number MFCD07772840
Chemical Name or Material N-Methyl-N-[(4-methyl-4H-thieno[3, 2-b]pyrrol-5-yl)methyl]amine hydrochloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC40996DA SDS Fisher Scientific Company
CC40996DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

335033-68-6
100.0
C9H12N2S·ClH
MFCD07772840
95%
94.5
95%
216.73
N-Methyl-N-[(4-methyl-4H-thieno[3, 2-b]pyrrol-5-yl)methyl]amine hydrochloride

Specifications

216.73
100.0
95%
94.5
335033-68-6
95%
1g
MFCD07772840
C9H12N2S·ClH
Amber Glass Bottle
N-Methyl-N-[(4-methyl-4H-thieno[3, 2-b]pyrrol-5-yl)methyl]amine hydrochloride

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning