missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-N-(3-thien-2-ylbenzyl)amine, 97%, Maybridge

Chemical Identifiers

CAS 859833-20-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H13NS
Formula Weight 203.31
MDL Number MFCD08435869
Chemical Name or Material N-Methyl-N-(3-thien-2-ylbenzyl)amine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC44546CB SDS Fisher Scientific Company
CC44546CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC44546DA SDS Fisher Scientific Company
CC44546DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

859833-20-8
100.0
C12H13NS
MFCD08435869
97%
96.5
97%
203.31
N-Methyl-N-(3-thien-2-ylbenzyl)amine

Specifications

203.31
100.0
Amber Glass Bottle
96.5
97%
97%
250mg
MFCD08435869
859833-20-8
C12H13NS
N-Methyl-N-(3-thien-2-ylbenzyl)amine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger