Please login to your online account to display your discounted pricing

N-Methyl-N-(3-morpholin-4-ylbenzyl)amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material N-methyl-N-(3-morpholin-4-ylbenzyl)amine
CAS 864068-83-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H18N2O
Formula Weight 206.29
MDL Number MFCD07772813
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC33846CB

  SDS
fisher scientific company
CC33846CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC33846DA

  SDS
fisher scientific company
CC33846DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.29
Quantity 250mg
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
MDL Number MFCD07772813
Assay Percent Range 97%
CAS Min % 96.5
Chemical Name or Material N-methyl-N-(3-morpholin-4-ylbenzyl)amine
Molecular Formula C12H18N2O
CAS 864068-83-7
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.